Learn More
LNI Swissgas NG CASTORE BASIC 25.40
Product Code. 17802059 Shop All LNI Swissgas Products
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. 17802059
Brand: LNI Swissgas 6920.52.026
Nitrogen generator with air outlet and no dryer.
  • Needs source of dry and oil free air.
Specifications
Voltage 100/115/220/230 V
Type Nitrogen Gas Generator