missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NG2/MCSP Antibody (7.1), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB25851H
This item is not returnable.
View return policy
Description
NG2/MCSP Monoclonal antibody specifically detects NG2/MCSP in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF
Specifications
| NG2/MCSP | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 1464 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunoprecipitation, CyTOF | |
| 7.1 | |
| Flow Cytometry, Immunoprecipitation, CyTOF-ready | |
| chondroitin sulfate proteoglycan 4, chondroitin sulfate proteoglycan 4 (melanoma-associated), EC 3.6.3, HMW-MAA, MCSPChondroitin sulfate proteoglycan NG2, MCSPGMelanoma chondroitin sulfate proteoglycan, MEL-CSPG, MSK16Melanoma-associated chondroitin sulfate proteoglycan, NG2EC 2.7.8 | |
| Human transformed stromal cells | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction