missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NGAL Monoclonal Antibody (8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ ABS0390802
This item is not returnable.
View return policy
Description
ABS 039-08-02 detects NGAL in rat samples. This antibody will not cross react with human, pig, dog, or mouse NGAL. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
NGAL (Neutrophil gelatinase-associated lipocalin, lipocalin 2, siderocalin and neutrophil lipocalin) is a 25 kDa glycosylated single chain monomer and member of the lipocalin family of proteins that bind and transport small lipophilic molecules. NGAL is released by activated neutrophils, can form dimers and small amounts of higher oligomers, and complexes with matrix metalloproteinase 9 (MMP-9, gelatinase B). Low level expression of NGAL in a variety of epithelia may be increased in inflammation or cancers. Expression of NGAL in the kidney is dramatically increased by ischemic or nephrotoxic kidney injury.
Specifications
| NGAL | |
| Monoclonal | |
| 1.01 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P30152 | |
| LCN2 | |
| Purified recombinant rat NGAL precursor. | |
| 400 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| 8 | |
| Unconjugated | |
| LCN2 | |
| 24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; alpha-2-microglobulin-related protein; Alpha-2U globulin-related protein; AW212229; HGNC:6526; HNL; LCN2; lipocalin 2; lipocalin 2 (oncogene 24p3); Lipocalin 24p3; Lipocalin2; Lipocalin-2; migration-stimulating factor inhibitor; MSFI; neutrophil gelatinase-associated lipocalin; NGAL; oncogene 24p3; OTTHUMP00000022240; p25; RP23-161B9.11; secreted inducible protein 24; Siderocalin; Siderocalin LCN2; Sip24; SV-40-induced 24P3 protein; uterocalin | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 170496 | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction