missing translation for 'onlineSavingsMsg'
Learn More

NGDN Monoclonal Antibody (OTI1B8), TrueMAB™, OriGene

Product Code. 17520484
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17520484 100 μL 100µL
17540484 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17520484 Supplier Origene Technologies Supplier No. TA808822

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Involved in the translational repression of cytoplasmic polyadenylation element (CPE)-containing mRNAs.

Specifications

Antigen NGDN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NGDN
Gene Accession No. Q8NEJ9
Gene Alias C14orf120, CANu1, LCP5, NGD, lpd-2
Gene Symbols NGDN
Host Species Mouse
Immunogen Full length human recombit protein of human NGDN produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25983
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.