missing translation for 'onlineSavingsMsg'
Learn More

NGDN Monoclonal Antibody (OTI1B8), TrueMAB™, OriGene

Product Code. 17540484
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17540484 30 μL 30µL
17520484 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17540484 Supplier Origene Technologies Supplier No. TA808822S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NGDN Monoclonal antibody specifically detects NGDN in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen NGDN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene UBP1
Gene Accession No. Q9NZI7
Gene Alias 1500001L15Rik; C14orf120; CANu1; Centromere accumulated nuclear protein 1; EIF4E-binding protein; eukaryotic initiation factor 4E and CPEB binding protein; LCP5; lpd-2; neuroguidin; neuroguidin, EIF4E binding protein; NGD; Ngdn; RGD1310211
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 47-269 of human UBP1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7342
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.