missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NGDN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587151
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
Involved in the translational repression of cytoplasmic polyadenylation element (CPE)-containing mRNAs.
Specifications
| NGDN | |
| Polyclonal | |
| Unconjugated | |
| Ngdn | |
| 1500001L15Rik; C14orf120; CANu1; Centromere accumulated nuclear protein 1; EIF4E-binding protein; eukaryotic initiation factor 4E and CPEB binding protein; LCP5; lpd-2; neuroguidin; neuroguidin, EIF4E binding protein; NGD; Ngdn; RGD1310211 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25983, 305887, 68966 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NEJ9, Q9DB96 | |
| Ngdn | |
| Recombinant fusion protein of human NGDN. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction