Learn More
Invitrogen™ NGF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589685
Description
Positive Samples: mouse heart, mouse brain Immunogen sequence: EPHSESNVPA GHTIPQAHWT KLQHSLDTAL RRARSAPAAA IAARVAGQTR NITVDPRLFK KRRLRSPRVL FSTQPPREAA DTQDLDFEVG GAAPFNRTHR SKRSSSHPIF HRGEFSVCDS VSVWVGDKTT ATDIKGKEVM VLGEVNINNS VFKQYFFETK CRDPNPVDSG CRGIDSKHWN SYCTTTHTFV KALTMDGKQA AWRFIRIDTA CVCVLSRKAV RRA.
Nerve growth factor (NGF) influences the survival and differentiation of a variety of neuronal and nonneuronal cells. The actions of this polypeptide result from binding to specific cell surface receptors which are present as both high and low affinity sites. Although both types of receptor bind NGF only the high affinity form is capable of signal transduction. Cross-linking studies with 125I-NGF have shown that the high affinity receptor of rat PC12 cells forms an Mr 158,000 complex with NGF while the low affinity receptor forms an Mr 100,000 complex. Full length cDNA clones for both the human and rat NGF receptor genes have been produced and sequenced, revealing greater than 90% homology between the two species. NGF exhibits nerve growth stimulating activity and is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Further, NGF stimulates division and differentiation of sympathetic and embryonic sensory neurons. Mutations in the NGF gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NGF gene expression is associated with allergic rhinitis.
Specifications
| NGF | |
| Polyclonal | |
| Unconjugated | |
| NGF | |
| B nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; Nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta polypeptide; nerve growth factor, beta subunit; NGF; NGF b; NGF β; Ngfb; NGFbeta; NGFβ; β nerve growth factor; β NGF; βNGF | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18049, 310738, 4803 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Western Blot | |
| 3.6 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P01138, P01139, P25427 | |
| NGF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 19-241 of human NGF (NP_0024972). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.