missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGF-Receptor / CD271 Mouse, Clone: NGFR5 + NTR/912, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4804-MSM3-P1
This item is not returnable.
View return policy
Specifications
| NGF-Receptor / CD271 | |
| Cell surface, cytoplasmic | |
| NGFR5 + NTR/912 | |
| Unconjugated | |
| NP_002498 | |
| Mouse | |
| 75kDa | |
| Protein A/G Purified | |
| RUO | |
| 4804 | |
| IgG |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| NGFR | |
| NGFR from A875 melanoma cells (NGFR5); Recombinant human p75 NGFR protein (NTR/912) | |
| Neuronal axons, Schwann cells,, perineural cells of peripheral nerves, or tumors of nerve sheath differentiation e.g. Schwannoma, Neurofibroma. Soma, axons of sensory neurons,, ganglionic satellite cells. Melanomas. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction