missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFI-B alpha/Nur77/NR4A1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56745AF532
This item is not returnable.
View return policy
Description
NGFI-B alpha/Nur77/NR4A1 Polyclonal specifically detects NGFI-B alpha/Nur77/NR4A1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| NGFI-B alpha/Nur77/NR4A1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Early response protein NAK1, GFRP1ST-59, growth factor-inducible nuclear protein N10, HMRNP10, hormone receptor, MGC9485, N10, NAK1, NAK-1, NGFIB, Nuclear hormone receptor NUR/77, nuclear receptor subfamily 4 group A member 1, nuclear receptor subfamily 4, group A, member 1, NUR77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, steroid receptor TR3, Testicular receptor 3, TR3, TR3 orphan receptor | |
| Rabbit | |
| Protein G purified | |
| Apoptosis | |
| 3164.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NR4A1 | |
| This antibody was developed against a synthetic peptide corresponding to aa 251-266 of human Nak1 (NP_775180). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction