Learn More
NGFR Mouse anti-Human, Clone: NGFR5, Abnova™
Mouse monoclonal antibody raised against native human NGFR.
Brand: Abnova MAB13385.100ug
Description
Nerve growth factor receptor contains an extracellular domain containing four 40-amino acid repeats with 6 cysteine residues at conserved positions followed by a serine/threonine-rich region, a single transmembrane domain, and a 155-amino acid cytoplasmic domain. The cysteine-rich region contains the nerve growth factor binding domain. [provided by RefSeq]
Specifications
| NGFR | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CD271/Gp80-LNGFR/TNFRSF16/p75(NTR)/p75NTR | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 4804 | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| NGFR5 | |
| Mouse monoclonal antibody raised against native human NGFR. | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
| NGFR | |
| A875 melanoma cells. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.