missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFR/TNFRSF16/p75NTR Antibody (SPM299), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11470PCP
This item is not returnable.
View return policy
Description
NGFR/TNFRSF16/p75NTR Monoclonal antibody specifically detects NGFR/TNFRSF16/p75NTR in Human, Feline, Ferret, Monkey, Baboon, Rabbit, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry
Specifications
| NGFR/TNFRSF16/p75NTR | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Feline, Ferret, Monkey, Baboon, Rabbit, Mouse (Negative), Rat (Negative) | |
| Purified |
| Flow Cytometry | |
| SPM299 | |
| Flow Cytometry | |
| CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF receptor, p75 ICD, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16 | |
| NGFR from A875 melanoma cells | |
| 0.1 mL | |
| Apoptosis, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| 4804 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction