missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NGLY1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29942
This item is not returnable.
View return policy
Description
NGLY1 Polyclonal antibody specifically detects NGLY1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| NGLY1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| EC 3.5.1.52, hPNGase, N-glycanase 1FLJ12409, Peptide:N-glycanase, peptide-N(4)-(N-acetyl-beta-glucosaminyl)asparagine amidase, PNG1FLJ11005, PNGase | |
| Between 250 and 300 | |
| 100 μg | |
| Cell Biology | |
| 55768 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction