missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: ISDEDFLLLE LLHWFKEEFF HWVNNVLCSK CGGQTRSRDR SLLPSDDELK WGAKEVEDHY CDACQFSNRF PRYNNPEKLL ETRCGR Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
This gene encodes an enzyme that catalyzes hydrolysis of an N(4)-(acetyl-beta-D-glucosaminyl) asparagine residue to N-acetyl-beta-D-glucosaminylamine and a peptide containing an aspartate residue. The encoded enzyme may play a role in the proteasome-mediated degradation of misfolded glycoproteins. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Antigen | NGLY1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NGLY1 |
| Gene Accession No. | Q96IV0 |
| Gene Alias | 1110002C09Rik; CDDG; CDG1V; hPNGase; mPNGase; NGLY1; N-glycanase 1; peptide N-glycanase homolog; Peptide:N-glycanase; peptide-N(4)-(N-acetyl-beta-glucosaminyl)asparagine amidase; PNG1; PNGase |
| Gene Symbols | NGLY1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?