missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NgR2/NgRH1/RTN4RL2 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41200C
This item is not returnable.
View return policy
Description
NgR2/NgRH1/RTN4RL2 Polyclonal antibody specifically detects NgR2/NgRH1/RTN4RL2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| NgR2/NgRH1/RTN4RL2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| DyLight 650 | |
| NgR2, NGRH1, reticulon 4 receptor-like 2 | |
| Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human RTN4RL2. The immunogen is located within amino acids 330 - 380 of RTN4RL2. Amino Acid Squence: LYRDLPAEDSRGRQGGDA | |
| 0.1 mL | |
| Cellular Signaling | |
| 349667 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction