missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGX6 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98040F
This item is not returnable.
View return policy
Description
NGX6 Polyclonal specifically detects NGX6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| NGX6 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| C9orf127, chromosome 9 open reading frame 127, NAG-5, nasopharyngeal carcinoma expressed 6, nasopharyngeal carcinoma related protein, Nasopharyngeal carcinoma-associated gene 6 protein, NGX6MGC120460, Protein NAG-5, Protein NGX6, RP11-112J3.10, transmembrane protein 8B | |
| Produced in rabbits immunized with E. coli-derived Human NGX6 fragment. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 51754 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| FITC | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction