missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE8 Antibody (7A11), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-62091G
This item is not returnable.
View return policy
Description
NHE8 Monoclonal specifically detects NHE8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| NHE8 | |
| Monoclonal | |
| DyLight 488 | |
| Q9Y2E8 | |
| SLC9A8 | |
| A fusion protein containing the C-terminal 89 residues of human NHE8. [Swiss-Prot# Q9Y2E8] | |
| Protein G purified | |
| RUO | |
| Primary | |
| This is specific for the 85 kDa mature glycosylated form of the protein. It does not recognize the 55 kDa immature form. | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 7A11 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| DKFZp686C03237, KIAA0939DKFZp686C03237, MGC138418, Na(+)/H(+) exchanger 8, NHE-8, NHE8FLJ42500, sodium/hydrogen exchanger 8, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 8, solute carrier family 9 (sodium/hydrogen exchanger), member 8, Solute carrier family 9 member 8 | |
| Mouse | |
| 80 kDa | |
| 0.1 mL | |
| Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism | |
| 23315 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction