missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHEJ1 Monoclonal Antibody (OTI3B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502002
This item is not returnable.
View return policy
Description
Double-strand breaks in DNA result from genotoxic stresses and are among the most damaging of DNA lesions. This gene encodes a DNA repair factor essential for the nonhomologous end-joining pathway, which preferentially mediates repair of double-stranded breaks. Mutations in this gene cause different kinds of severe combined immunodeficiency disorders.Specifications
| NHEJ1 | |
| Monoclonal | |
| 0.71 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H9Q4 | |
| NHEJ1 | |
| Full length human recombit protein of human NHEJ1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI3B5 | |
| Unconjugated | |
| NHEJ1 | |
| XLF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79840 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction