missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NHERF2 (Isoform B/E) Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142185
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize isoforms b and e (NP_004776.3 and NP_001239005.1) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This gene encodes a member of the NHERF family of PDZ scaffolding proteins. These proteins mediate many cellular processes by binding to and regulating the membrane expression and protein-protein interactions of membrane receptors and transport proteins. The encoded protein plays a role in intestinal sodium absorption by regulating the activity of the sodium/hydrogen exchanger 3, and may also regulate the cystic fibrosis transmembrane regulator (CFTR) ion channel. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Nov 2011].
Specifications
| NHERF2 (Isoform B/E) | |
| Polyclonal | |
| Unconjugated | |
| SLC9A3R2 | |
| SLC9A3R2 | |
| Peptide with sequence C-DLPGSDKDTEESGLH. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q15599 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 9351 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction