missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHLH2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98163F
This item is not returnable.
View return policy
Description
NHLH2 Polyclonal antibody specifically detects NHLH2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NHLH2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| FITC | |
| BHLHA34, bHLHa34NSCL-2, Class A basic helix-loop-helix protein 34, helix-loop-helix protein 2, HEN-2, HEN2NSCL2, nescient helix loop helix 2KIAA0490 | |
| Produced in rabbits immunized with E. coli-derived Human NHLH2 fragment. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 4808 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction