missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NHLRC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17191124 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17191124 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17191124 Supplier Invitrogen™ Supplier No. PA5113268

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NHLRC2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

NHLRC2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NHLRC2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NHLRC2
Gene Accession No. Q8NBF2
Gene Alias 1200003G01Rik; AI835049; AV002846; AW496455; NHL repeat containing 2; NHL repeat-containing protein 2; Nhlrc2; novel NHL repeat domain containing protein
Gene Symbols NHLRC2
Host Species Rabbit
Immunogen Recombinant Human NHL repeat-containing protein 2 protein (1-367AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374354
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.