missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHP2L1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93592-0.02ml
This item is not returnable.
View return policy
Description
NHP2L1 Polyclonal antibody specifically detects NHP2L1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| NHP2L1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| [U4/U6.U5] tri-snRNP 15.5 kD RNA binding protein, 15.5K, FA1, FA-1, High mobility group-like nuclear protein 2 homolog 1, hSNU13, NHP2 non-histone chromosome protein 2-like 1 (S. cerevisiae), NHP2-like protein 1, NHPX, non-histone chromosome protein 2 (S. cerevisiae)-like 1, non-histone chromosome protein 2-like 1, OTK27, small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5), SNRNP15-5, SNU13, SNU13 homolog, SPAG12, sperm specific antigen 1, SSFA1, U4/U6.U5 tri-snRNP 15.5 kDa protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-128 of human NHP2L1 (NP_004999.1). MTEADVNPKAYPLADAHLTKKLLDLVQQSCNYKQLRKGANEATKTLNRGISEFIVMAADAEPLEIILHLPLLCEDKNVPYVFVRSKQALGRACGVSRPVIACSVTIKEGSQLKQQIQSIQQSIERLLV | |
| 0.02 mL | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| 4809 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction