Learn More
NHP2L1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15802-1-AP-20UL
This item is not returnable.
View return policy
Description
Originally named because of its sequence similarity to the Saccharomyces cerevisiae NHP2 (non-histone protein 2), this protein appears to be a highly conserved nuclear protein that is a component of the [U4/U6.U5] tri-snRNP. It binds to the 5′ stem-loop of U4 snRNA. Two transcript variants encoding the same protein have been found for this gene.Specifications
| NHP2L1 | |
| Polyclonal | |
| Unconjugated | |
| SNU13 | |
| 15.5K, FA 1, FA1, hSNU13, NHP2 like protein 1, NHP2L1, NHPX, OTK27, SNRNP15 5, SNU13, SNU13 homolog, SPAG12, SSFA1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20826, 4809 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P55769, Q9D0T1 | |
| SNU13 | |
| NHP2L1 Fusion Protein Ag8532 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |