missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NHS Polyclonal Antibody

Product Code. 15950075
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15950075 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15950075 Supplier Invitrogen™ Supplier No. PA567417

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ATYDSFLEKSP SDKADTSSHF SVDTEGYYTS MHFDCGLKGN KSYVCHYAAL GPENGQGVGA SPGLPDCAWQ DYLDHKRQGR PSISFRKP Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.

May function in cell morphology by maintaining the integrity of the circumferential actin ring and controlling lamellipod formation. Involved in the regulation eye, tooth, brain and craniofacial development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NHS
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NHS
Gene Accession No. Q6T4R5
Gene Alias congenital cataracts and dental anomalies protein; CTRCT40; CXN; Nance-Horan syndrome (congenital cataracts and dental anomalies); Nance-Horan syndrome protein; NHS; NHS actin remodeling regulator; SCML1
Gene Symbols NHS
Host Species Rabbit
Immunogen Recombinant Human NHS. Recombinant protein control fragment (Product #RP-108078).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4810
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.