missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niban Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 572.00
Specifications
| Antigen | Niban |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18463821
|
Novus Biologicals
NBP1-88340 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18404971
|
Novus Biologicals
NBP1-88340-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Niban Polyclonal antibody specifically detects Niban in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Niban | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| C1orf24, cell growth inhibiting protein 39, Cell growth-inhibiting gene 39 protein, chromosome 1 open reading frame 24, family with sequence similarity 129, member A, GIG39, NIBANFLJ38228, Protein FAM129A, protein Niban | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VESYENKEAYQRGAAPKCRILPAGGKVLTSEDEYNLLSDRHFPDPLASSEKENTQPFVVLPKEFPVYLWQPFFRHGYFCFHEAADQKRFSALLS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 116496 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title