Learn More
Nicastrin Antibody (004), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89826PCP
Product Code. 18696401
  • ONLINE EXCLUSIVE PRICE
    € 641.00
    € 605.85 / 0.10mL
    Save € 35.15
    5% Off
Description
Nicastrin Monoclonal antibody specifically detects Nicastrin in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nicastrin
Monoclonal
PerCP
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Immunocytochemistry
004
Immunocytochemistry/ Immunofluorescence
anterior pharynx-defective 2, ATAG1874, KIAA0253APH2, nicastrin
This antibody was obtained from a rabbit immunized with purified, recombinant Human Nicastrin (Accession#: NP_056146.1; Met1-Glu669).
0.1 mL
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
23385
Store at 4C in the dark.