missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicastrin Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77269MFV500
This item is not returnable.
View return policy
Description
Nicastrin Polyclonal antibody specifically detects Nicastrin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Nicastrin | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| anterior pharynx-defective 2, ATAG1874, KIAA0253APH2, nicastrin | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human Nicastrin. The immunogen is located within amino acids 630 - 680 of Nicastrin. Amino Acid Squence: TEYSTWTESRWKDIRAR | |
| 0.1 mL | |
| Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 23385 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction