Learn More
NICE-3 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-97389AF647
Code produit. 18514403
  • € 415.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
NICE-3 Polyclonal antibody specifically detects NICE-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
NICE-3
Polyclonal
50mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
25912
Store at 4°C in the dark.
IgG
Immunohistochemistry (Paraffin)
Alexa Fluor 647
chromosome 1 open reading frame 43, HCV NS5A-transactivated protein 4
Produced in rabbits immunized with E. coli-derived Human NICE-3 fragment.
0.1 mL