Learn More
Nickel Standard Solution Concentrate, 10ppm, Reagecon™
BIOPHARMA
Product Code. 11434769 Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 11434769
Brand: Reagecon 5002000C