Learn More
Nickel Standard Solution (Plasma HIQU), Chem Lab™
Product Code. 11929102
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 11929102
Brand: Chem-Lab CL01.1421.0100
Specifications
Concentration 1000 μg/mL
Quantity 100 mL
compliance-icons
Product Identifier
  • Nickel standard solution 1000
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Nitric acid 69%