Learn More
β-Nicotinamide Mononucleotide Mouse, anti-Human, Clone: 3A1F11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68638-1-IG-20UL
 View more versions of this product
Product Code. 18068955
  • € 145.00 / 20µL
Description
The β-Nicotinamide Mononucleotide antibody from Proteintech is a mouse monoclonal antibody generated with ptg of na β-Nicotinamide Mononucleotide. This antibody recognizes human antigen. The β-Nicotinamide Mononucleotide antibody has been validated for the following applications: ELISA analysis.
Specifications
Specifications
β-Nicotinamide Mononucleotide
3A1F11
NMN, β NMN, β-Nicotinamide Mononucleotide, β-NMN
20 μL
Human
150 kDa
IgG1
ELISA
1000 μg/mL
Mouse
Primary
Store at -20°C. Stable for one year after shipment.