Learn More
β-Nicotinamide Mononucleotide Mouse, anti-Human, Clone: 3A1F11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68638-1-IG-150UL
Description
The β-Nicotinamide Mononucleotide antibody from Proteintech is a mouse monoclonal antibody generated with ptg of na β-Nicotinamide Mononucleotide. This antibody recognizes human antigen. The β-Nicotinamide Mononucleotide antibody has been validated for the following applications: ELISA analysis.
Specifications
| β-Nicotinamide Mononucleotide | |
| 3A1F11 | |
| NMN, β NMN, β-Nicotinamide Mononucleotide, β-NMN | |
| 150 μL | |
| Human | |
| 150 kDa | |
| IgG1 |
| ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |