missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinamide N-Methyltransferase/NNMT Antibody (AT11G11), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50581AF350
This item is not returnable.
View return policy
Description
Nicotinamide N-Methyltransferase/NNMT Monoclonal specifically detects Nicotinamide N-Methyltransferase/NNMT in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Nicotinamide N-Methyltransferase/NNMT | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NNMT | |
| Recombinant human NNMT (1-264aa) purified from E. coli | |
| Primary | |
| Human | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| AT11G11 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| EC 2.1.1.1, nicotinamide N-methyltransferase | |
| Mouse | |
| 100 μL | |
| 4837 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction