missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinamide N-Methyltransferase/NNMT Antibody (AT11G11), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50581PECY7
This item is not returnable.
View return policy
Description
Nicotinamide N-Methyltransferase/NNMT Monoclonal specifically detects Nicotinamide N-Methyltransferase/NNMT in Human samples. It is validated for Flow Cytometry.
Specifications
| Nicotinamide N-Methyltransferase/NNMT | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| NNMT | |
| Recombinant human NNMT (1-264aa) purified from E. coli | |
| 0.1 mL | |
| 4837.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| AT11G11 | |
| Flow Cytometry | |
| EC 2.1.1.1, nicotinamide N-methyltransferase | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Human | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction