Learn More
Nicotinic Acetylcholine R alpha 1/CHRNA1 Antibody (mAb 192), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-81070PCP
Product Code. 18683711
  • € 570.00 / 0.01mL
Description
Nicotinic Acetylcholine R alpha 1/CHRNA1 Monoclonal antibody specifically detects Nicotinic Acetylcholine R alpha 1/CHRNA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Neutralization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunocytochemistry
PerCP
Primary
Rat
Purified
Monoclonal
RUO
1134
Store at 4C in the dark.