missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 4/CHRNA4 Antibody (SR2141), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22215-50ul
This item is not returnable.
View return policy
Description
Nicotinic Acetylcholine R alpha 4/CHRNA4 Monoclonal antibody specifically detects Nicotinic Acetylcholine R alpha 4/CHRNA4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Nicotinic Acetylcholine R alpha 4/CHRNA4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR2141 | |
| Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 | |
| BFNC, cholinergic receptor, nicotinic, alpha 4, cholinergic receptor, nicotinic, alpha polypeptide 4, EBN, EBN1, ENFL1, FLJ95812, NACHR, NACHRA4, NACRA4, neuronal acetylcholine receptor subunit alpha-4, neuronal nicotinic acetylcholine receptor alpha-4 subunit | |
| A synthesized peptide derived from human Nicotinic Acetylcholine R alpha 4/CHRNA4 (Uniprot #: P43681) | |
| 50 μg | |
| Neuroscience | |
| 1137 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction