missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 7/CHRNA7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 214.00 - € 584.00
Specifications
| Antigen | Nicotinic Acetylcholine R alpha 7/CHRNA7 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30226846
|
Novus Biologicals
NBP3-35614-100ul |
100 μL |
€ 584.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232980
|
Novus Biologicals
NBP3-35614-20ul |
20 μL |
€ 214.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nicotinic Acetylcholine R alpha 7/CHRNA7 Polyclonal antibody specifically detects Nicotinic Acetylcholine R alpha 7/CHRNA7 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ImmunofluorescenceSpecifications
| Nicotinic Acetylcholine R alpha 7/CHRNA7 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 1139 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| a7 nicotinic acetylcholine receptor, alpha 7 neuronal nicotinic acetylcholine receptor, alpha-7 nicotinic cholinergic receptor subunit, cholinergic receptor, nicotinic, alpha 7, cholinergic receptor, nicotinic, alpha polypeptide 7, CHRNA7-2, NACHRA7, neuronal acetylcholine receptor protein, alpha-7 chain, neuronal acetylcholine receptor subunit alpha-7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 23-230 of human Nicotinic Acetylcholine R alpha 7/CHRNA7 (NP_001177384.1).,, Sequence:, ASPPSTPPWDPGHIPGASVRPAPGPVSLQGEFQRKLYKELVKNYNPLERPVANDSQPLTVYFSLSLLQIMDVDEKNQVLTTNIWLQMSWTDHYLQWNVSEYPGVKTVRFPDGQIWKPDILLYNSADERFDATFHTNVLVNSSGHCQYLPPGIFKSSCYIDVRWFPFDVQHCKLKFGSWSYGGWSLDLQMQEADISGYIPNGEWDLVGI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto