missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine Receptor beta 4 Antibody (1H11F2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61743
This item is not returnable.
View return policy
Description
Nicotinic Acetylcholine Receptor beta 4 Monoclonal specifically detects Nicotinic Acetylcholine Receptor beta 4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Nicotinic Acetylcholine Receptor beta 4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
| cholinergic receptor, nicotinic, beta 4, cholinergic receptor, nicotinic, beta polypeptide 4, neuronal acetylcholine receptor subunit beta-4, neuronal nicotinic receptor beta 4 subunit | |
| Mouse | |
| 56.4 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1H11F2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CHRNB4 | |
| Purified recombinant fragment of human CHRNB4 (AA: extra 22-236) expressed in E. Coli. | |
| Protein G purified | |
| Cellular Markers, Neuronal Cell Markers | |
| 1143 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction