missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine Receptor epsilon Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 201.00 - € 470.00
Specifications
| Antigen | Nicotinic Acetylcholine Receptor epsilon |
|---|---|
| Dilution | Western Blot 1:1000-1:4000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656991
|
Novus Biologicals
NBP2-94553-0.02ml |
0.02 mL |
€ 201.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670161
|
Novus Biologicals
NBP2-94553-0.1ml |
0.1 mL |
€ 470.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nicotinic Acetylcholine Receptor epsilon Polyclonal antibody specifically detects Nicotinic Acetylcholine Receptor epsilon in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Nicotinic Acetylcholine Receptor epsilon | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 1145 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:4000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| acetylcholine receptor subunit epsilon, AchR epsilon subunit, ACHREFCCMS, cholinergic receptor, nicotinic, epsilon, cholinergic receptor, nicotinic, epsilon polypeptide, CMS1D, CMS1E, CMS2A, SCCMS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 21-239 of human CHRNE (NP_000071.1). KNEELRLYHHLFNNYDPGSRPVREPEDTVTISLKVTLTNLISLNEKEETLTTSVWIGIDWQDYRLNYSKDDFGGIETLRVPSELVWLPEIVLENNIDGQFGVAYDANVLVYEGGSVTWLPPAIYRSVCAVEVTYFPFDWQNCSLIFRSQTYNAEEVEFTFAVDNDGKTINKIDIDTEAYTENGEWAIDFCPGVIRRHHGGATDGPGETDVIYSLIIRRK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title