missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nidogen-1/Entactin Antibody (ELM1), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-97701R
This item is not returnable.
View return policy
Description
Nidogen-1/Entactin Monoclonal antibody specifically detects Nidogen-1/Entactin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| Nidogen-1/Entactin | |
| Monoclonal | |
| DyLight 550 | |
| enactin, Entactin, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1 | |
| Derived by fusion of X63 Ag 8.653 mouse myeloma cells with spleen cells from a Fisher rat immunized with a partially purified preparation of laminin from the EHS mouse tumor. | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen) | |
| ELM1 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 4811 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction