missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nidogen 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145302
This item is not returnable.
View return policy
Description
Basement membranes, which are composed of type IV collagens, laminins, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis. Basement membranes, which are composed of type IV collagens, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.
Specifications
| Nidogen 2 | |
| Polyclonal | |
| Unconjugated | |
| NID2 | |
| AW547149; entactin 2; entactin-2; LOW QUALITY PROTEIN: nidogen-2; Ly111; Nid2; NID-2; Nidogen; nidogen 2; nidogen 2 (osteonidogen); nidogen-2; nidogen-2; LOW QUALITY PROTEIN: nidogen-2; osteonidogen | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 22795 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q14112 | |
| NID2 | |
| Recombinant Human NID2 protein, Leu31-Ala289 (Accession #Q14112). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction