missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nidogen Polyclonal Antibody
GREENER_CHOICE

Product Code. 16539851
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16539851 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16539851 Supplier Invitrogen™ Supplier No. PA597016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DLDTTDGLGK VYYREDLSPS ITQRAAECVH RGFPEISFQP SSAVVVTWES VAPYQGPSRD PDQKGKRNTF QAVLASSDSS SYAIFLYPED GLQFHTTFSK KENNQVPAVV AFSQGSVGFL WKSNGAYNIF ANDRESVENL AKSSNSGQQG VWVFEIGSPA TTNGVVPADV ILGTEDGAEY DDEDEDYDLA TTRLGLEDVG TTPFSYKALR RGGADTYSVP SVLSPRRAAT ERPLGPPTER TRSFQLAVET FHQQHPQVID VDEVEETGVV FSYNTDSRQT CANNRHQCSV H; Positive Samples: LO2, 293T; Cellular Location: basement membrane, extracellular matrix, extracellular space.

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nidogen
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NID1
Gene Accession No. P14543
Gene Alias A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1;
Gene Symbols NID1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 110-400 of human NID1 (NP_0024992).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4811
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.