Learn More
Invitrogen™ Nidogen Recombinant Rabbit Monoclonal Antibody (24GB4995)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557119
This item is not returnable.
View return policy
Description
Nidogen Recombinant Monoclonal Antibody for Western Blot, Flow
This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
Specifications
| Nidogen | |
| Recombinant Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P08460, P10493 | |
| NID1 | |
| Recombinant protein of human Entactin/NID (aa 844-1245). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot | |
| 24GB4995 | |
| Unconjugated | |
| NID1 | |
| A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1; | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18073, 25494 | |
| -20°C | |
| Liquid |