missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NIF3L1 Monoclonal Antibody (OTI3E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503663
This item is not returnable.
View return policy
Description
ALS2CR1 may function as a transcriptional corepressor through its interaction with COPS2, negatively regulating the expression of genes involved in neuronal differentiation.Specifications
| NIF3L1 | |
| Monoclonal | |
| 0.22 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9GZT8 | |
| NIF3L1 | |
| Full length human recombit protein of human NIF3L1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3E1 | |
| Unconjugated | |
| NIF3L1 | |
| ALS2CR1, CALS-7, MDS015 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 60491 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction