Learn More
Tocris Bioscience™ Nimbolide
Description
Nimbolide inhibits auto-ubiquitination of the E3 ligase RNF114 and p21 ubiquitination in vitro. This compound impairs proliferation of triple-negative breast cancer cell lines. Nimbolide also induces apoptotic cell death in a Waldenströms macroglobulinemia cell line (BCWM1) and inhibits tumor growth in a xenograft model of Waldenströms macroglobulinemia.
Nimbolide can be used for the development of Degraders that utilize RNF114 as an E3 ligase for Targeted Protein Degradation.
Specifications
Specifications
| Chemical Name or Material | (2 aR,5 aR,6 S,6 aR,8 R,9 aR,10 AS,10 bR,10 cR)-8-(3-Furanyl)-2 A,5 A,6,6 A,8,9,9 A,10 A,10 b,10 c-decahydro-2 A,5 A,6 A,7-tetramethyl-2,5-dioxo-2 H,5 H-cyclopenta[d']naphtho[1,8-bc:2,3-b']difuran-6-acetic acid methyl ester |
| CAS | 25990-37-8 |
| Quantity | 5 mg |
| Target | Ubiquitin Ligase (E3) Inhibitors |
| Molecular Formula | C27H30O7 |
| Purity | 0.97 |