Learn More
Nimbolide, TRC
Specifications
Specifications
| Chemical Name or Material | Nimbolide |
| CAS | 25990-37-8 |
| Molecular Formula | C27H30O7 |
| Quantity | 10 mg |
| Synonym | (4α,5α,6α,7α,15β,17α)-7,15:21,23-Diepoxy-6-hydroxy-4,8-dimethyl-1-oxo-18,24-dinor-11,12-secochola-2,13,20,22-tetraene-4,11-dicarboxylic αcid γ-lactone Methyl Ester, NSC 309909 |
| InChI Formula | InChI=1S/C27H30O7/c1-13-15(14-7-9-32-12-14)10-16-20(13)27(4)17(11-19(29)31-5)26(3)18(28)6-8-25(2)22(26)21(23(27)33-16)34-24(25)30/h6-9,12,15-17,21-23H,10-11H2,1-5H3/t15-,16-,17-,21-,22+,23-,25-,26+,27-/m1/s1 |
| SMILES | COC(=O)CC1C2(C)C(OC3C(C=C(C)C23)c4cocc4)C5OC(=O)C6(C)C=CC(=O)C1(C)C56 |
| Molecular Weight (g/mol) | 466.52 |
| Formula Weight | 466.1992 |