missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ninein Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97595MFV500
This item is not returnable.
View return policy
Description
Ninein Polyclonal antibody specifically detects Ninein in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Ninein | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| Glycogen synthase kinase 3 beta-interacting protein, GSK3B-interacting protein, hNinein, KIAA1565, ninein, ninein (GSK3B interacting protein), ninein centrosomal protein | |
| Produced in rabbits immunized with E. coli-derived Human Ninein fragment. | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 51199 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction