missing translation for 'onlineSavingsMsg'
Learn More

Ningetinib Tosylate, MedChemExpress

Product Code. p-200097245
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490457

Brand: Medchem Express HY107145

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ningetinib Tosylate is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and c-Met, VEGFR2 and Axl, respectively.

Ningetinib Tosylate is a potent, orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6.7, 1.9 and c-Met, VEGFR2 and Axl, respectively.

Specifications

Chemical Name or Material Ningetinib Tosylate
CAS 1394820-77-9
Color White
Purity Grade Notes Research
Molecular Formula C38H37FN4O8S
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (45.73 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=CC=C(S(=O)(O)=O)C=C1.CC2=C(C(NC3=CC(F)=C(OC4=CC=NC5=C4C=CC(OCC(C)(O)C)=C5)C=C3)=O)C(N(C6=CC=CC=C6)N2C)=O
Molecular Weight (g/mol) 728.79
Formula Weight 728.79
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.