Learn More
NINJ2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14085-1-AP-20UL
This item is not returnable.
View return policy
Description
Homophilic cell adhesion molecule that promotes axonal growth. May play a role in nerve regeneration and in the formation and function of other tissues.Specifications
| NINJ2 | |
| Polyclonal | |
| Unconjugated | |
| NINJ2 | |
| Nerve injury induced protein 2, NINJ2, ninjurin 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29862, 4815, 59115 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JHE8, Q9JL89, Q9NZG7 | |
| Ninj2 | |
| NINJ2 Fusion Protein Ag5215 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |