missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ninjurin-1 Antibody (758926), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB51051D
This item is not returnable.
View return policy
Description
Ninjurin-1 Monoclonal antibody specifically detects Ninjurin-1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| Ninjurin-1 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4814 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 758926 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| Nerve injury-induced protein 1, nerve injury-induced protein-1, NIN1, NINJURIN, ninjurin 1, ninjurin-1 | |
| E. coli-derived recombinant human Ninjurin-1, Asp2-Val81, Accession # Q92982 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction