missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ninjurin 1 Monoclonal Antibody (758926)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524365
This item is not returnable.
View return policy
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Homophilic cell adhesion molecule that promotes axonal growth. May play a role in nerve regeneration and in the formation and function of other tissues. Cell adhesion requires divalent cations.
Specifications
| Ninjurin 1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q92982 | |
| Ninj1 | |
| E. coli-derived recombinant human Ninjurin-1 Asp2-Val81. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 758926 | |
| Unconjugated | |
| Ninj1 | |
| AU024536; Nerve injury-induced protein 1; nerve injury-induced protein-1; NIN1; Ninj1; NINJURIN; ninjurin 1; ninjurin-1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4814 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction